missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylpropanetrione 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 71°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 238.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Diphenylpropanetrione |
Safety and Handling
| TSCA | No |