missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylpropane 95.0+%, TCI America™
$242.33 - $242.33
Chemical Identifiers
| CAS | 1081-75-0 |
|---|---|
| Molecular Formula | C15H16 |
| Molecular Weight (g/mol) | 196.29 |
| MDL Number | MFCD00043574 |
| InChI Key | VEAFKIYNHVBNIP-UHFFFAOYSA-N |
| Synonym | 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci |
| PubChem CID | 14125 |
| ChEBI | CHEBI:34060 |
| IUPAC Name | (3-phenylpropyl)benzene |
| SMILES | C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Chemical Identifiers
| 1081-75-0 | |
| 196.29 | |
| VEAFKIYNHVBNIP-UHFFFAOYSA-N | |
| 14125 | |
| (3-phenylpropyl)benzene |
| C15H16 | |
| MFCD00043574 | |
| 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci | |
| CHEBI:34060 | |
| C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Specifications
| 1081-75-0 | |
| 83°C | |
| MFCD00043574 | |
| 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci | |
| C(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
| 196.29 | |
| CHEBI:34060 | |
| ≥95.0% (GC) | |
| 1,3-Diphenylpropane |
| Yellow | |
| C15H16 | |
| 5 mL | |
| VEAFKIYNHVBNIP-UHFFFAOYSA-N | |
| (3-phenylpropyl)benzene | |
| 14125 | |
| 196.29 | |
| Liquid |
Safety and Handling
TSCA : No