missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylpropane 95.0+%, TCI America™

Catalog No. D14125ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14125ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14125ML Supplier TCI America Supplier No. D14125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1081-75-0
Molecular Formula C15H16
Molecular Weight (g/mol) 196.29
MDL Number MFCD00043574
InChI Key VEAFKIYNHVBNIP-UHFFFAOYSA-N
Synonym 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci
PubChem CID 14125
ChEBI CHEBI:34060
IUPAC Name (3-phenylpropyl)benzene
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 mL
Formula Weight 196.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diphenylpropane