Learn More
1,3-Diphenylpropane, 98%
CAS: 1081-75-0 | C15H16 | 196.29 g/mol
Supplier: Thermo Scientific Chemicals L0884909
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 1,3-Diphenylpropane | |
| 0.98 | |
| C15H16 | |
| 1.56 | |
| 10 g | |
| 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci | |
| VEAFKIYNHVBNIP-UHFFFAOYSA-N | |
| 3-phenylpropylbenzene | |
| 14125 | |
| 196.29 |
| 1081-75-0 | |
| 158°C to 159°C (10 mmHg) | |
| C6H5(CH2)3C6H5 | |
| MFCD00043574 | |
| 2044541 | |
| Not miscible or difficult to mix with water. | |
| C(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
| 196.29 | |
| CHEBI:34060 | |
| 98% |
Chemical Identifiers
| 1081-75-0 | |
| 196.29 | |
| VEAFKIYNHVBNIP-UHFFFAOYSA-N | |
| 14125 | |
| 3-phenylpropylbenzene |
| C15H16 | |
| MFCD00043574 | |
| 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci | |
| CHEBI:34060 | |
| C(CC1=CC=CC=C1)CC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 214-101-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only