missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylpropane, 98%

Catalog No. AAL0884904
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0884904 2 g
AAL0884909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0884904 Supplier Thermo Scientific Chemicals Supplier No. L0884904
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1081-75-0 | C15H16 | 196.29 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1081-75-0
Molecular Formula C15H16
Molecular Weight (g/mol) 196.29
MDL Number MFCD00043574
InChI Key VEAFKIYNHVBNIP-UHFFFAOYSA-N
Synonym 1,3-diphenylpropane, 3-phenylpropyl benzene, dibenzylmethane, propane, 1,3-diphenyl, unii-x3diq9w9po, 3-phenyl-propyl-benzene, x3diq9w9po, benzene, 1,1'-1,3-propanediyl bis, propane, 1,3-diphenyl-8ci, benzene, 1,1'-1,3-propanediyl bis-9ci
PubChem CID 14125
ChEBI CHEBI:34060
IUPAC Name 3-phenylpropylbenzene
SMILES C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Density 0.98
Boiling Point 158°C to 159°C (10 mmHg)
Refractive Index 1.56
Linear Formula C6H5(CH2)3C6H5
Quantity 2 g
Beilstein 2044541
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 196.29
Percent Purity 98%
Chemical Name or Material 1,3-Diphenylpropane
EINECSNumber 214-101-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only