missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylisobenzofuran, 97%
CAS: 5471-63-6 | C20H14O | 270.32 g/mol
$70.30 - $70.30
Chemical Identifiers
| CAS | 5471-63-6 |
|---|---|
| Molecular Formula | C20H14O |
| Molecular Weight (g/mol) | 270.32 |
| MDL Number | MFCD00005931 |
| InChI Key | ZKSVYBRJSMBDMV-UHFFFAOYSA-N |
| Synonym | 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, # |
| PubChem CID | 21649 |
| IUPAC Name | 1,3-diphenyl-2-benzofuran |
| SMILES | C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC147580010
|
Thermo Scientific Chemicals
147580010 |
1 g | Glass bottle |
Each for $70.30
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
| 5471-63-6 | |
| 270.32 | |
| ZKSVYBRJSMBDMV-UHFFFAOYSA-N | |
| 21649 | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4 |
| C20H14O | |
| MFCD00005931 | |
| 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, # | |
| 1,3-diphenyl-2-benzofuran |
Specifications
| 5471-63-6 | |
| 100.0 | |
| Yellow to Green | |
| 96% min. (GC) | |
| C20H14O | |
| 1 g | |
| 01,342; 02,178; 04,209 | |
| ZKSVYBRJSMBDMV-UHFFFAOYSA-N | |
| 1,3-diphenyl-2-benzofuran | |
| 21649 | |
| 97% | |
| 1, 3-Diphenylisobenzofuran, 99+% |
| 96.0 | |
| 129.0°C to 133.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005931 | |
| 17, 87 | |
| 1,3-diphenylisobenzofuran, diphenylisobenzofuran, isobenzofuran, 1,3-diphenyl, 1,3 diphenylisobenzofuran, 2,5-diphenyl-3,4-benzofuran, acmc-209lj9, 1,3-diphenylbenzo c furan, 1,3-diphenylisobenzo b furan, 1,3-di phenyl-2-benzofuran, # | |
| C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4 | |
| 270.32 | |
| 270.32 | |
| Crystalline Solid or Needles |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 226-808-5
TSCA : TSCA
RUO – Research Use Only