missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenylguanidine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 147°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 211.27 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Diphenylguanidine |
Safety and Handling
| EINECSNumber | (3)-0480&(3)-2189 |
| RTECSNumber | MF0875000 |
| TSCA | Yes |