missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylguanidine 98.0+%, TCI America™

Catalog No. D089225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D089225G 25 g
D0892500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D089225G Supplier TCI America Supplier No. D089225G

Chemical Identifiers

CAS 102-06-7
Molecular Formula C13H13N3
Molecular Weight (g/mol) 211.268
MDL Number MFCD00001758
InChI Key OWRCNXZUPFZXOS-UHFFFAOYSA-N
Synonym 1,3-diphenylguanidine, diphenylguanidine, vulkazit, accelerator d, n,n'-diphenylguanidine, melaniline, denax, vulkacite d, nocceler d, vulkacit d
PubChem CID 7594
IUPAC Name 1,2-diphenylguanidine
SMILES C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)N

Specifications

Melting Point 147°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 211.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenylguanidine
EINECSNumber (3)-0480&(3)-2189
RTECSNumber MF0875000
TSCA Yes