missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylguanidine, 97%

Catalog No. AAA1523422
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1523422 100 g
AAA1523430 250 g
AAA152340B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1523422 Supplier Thermo Scientific Chemicals Supplier No. A1523422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-06-7 | C13H13N3 | 211.268 g/mol

1,3-Diphenylguanidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenylguanidine is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 102-06-7
Molecular Formula C13H13N3
Molecular Weight (g/mol) 211.268
MDL Number MFCD00001758
InChI Key OWRCNXZUPFZXOS-UHFFFAOYSA-N
Synonym 1,3-diphenylguanidine, diphenylguanidine, vulkazit, accelerator d, n,n'-diphenylguanidine, melaniline, denax, vulkacite d, nocceler d, vulkacit d
PubChem CID 7594
IUPAC Name 1,2-diphenylguanidine
SMILES C1=CC=C(C=C1)NC(=NC2=CC=CC=C2)N

Specifications

Melting Point 146°C to 150°C
Density 1.13
Boiling Point >170°C (decomposition)
Flash Point 170°C (338°F)
Odor Characteristic
Quantity 100 g
UN Number UN3077
Beilstein 1875653
Merck Index 14,3324
Solubility Information Soluble in alcohol, chloroform, hot benzene, hot toluene, and dilute mineral acids
Formula Weight 211.27
Percent Purity 97%
Chemical Name or Material 1,3-Diphenylguanidine
Show More Show Less
Hazard Category H302-H315-H319-H335-H361f
Hazard Statement GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-002-1
RTECSNumber MF0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only