missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenylacetone, 99%

Catalog No. p-4258981
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117170250 25 g Glass bottle
AC117171000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117170250 Supplier Thermo Scientific Chemicals Supplier No. 117170250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102-04-5 | C15H14O | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004795
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES O=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 30.0°C to 34.0°C
Color Yellow
Density 1.0400g/mL
Boiling Point 330.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2COCH2C6H5
Quantity 25 g
Beilstein 07,445
Fieser 01,198; 04,123
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble ethanol, diethyl ether, petroleum ether
Formula Weight 210.27
Percent Purity 99%
Physical Form Crystalline Mass or Liquid
Chemical Name or Material 1, 3-Diphenylacetone
Show More Show Less
EINECSNumber 203-000-

RUO – Research Use Only