missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-propyn-1-ol, tech. 90%

Catalog No. p-7035655
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5337403 1 g
AAH5337406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5337403 Supplier Thermo Scientific Chemicals Supplier No. H5337403

CAS: 1817-49-8 | C15H12O | 208.26 g/mol

1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-2-propyn-1-ol is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1817-49-8
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD06654198
InChI Key DZZWMODRWHHWFR-UHFFFAOYSA-N
Synonym 1,3-diphenyl-2-propyn-1-ol, 1,3-diphenyl-prop-2-yn-1-ol, 1-phenyl-3-phenyl-propyn-3-ol, 1,3-diphenylpropargyl alcohol, benzenemethanol, a-2-phenylethynyl, alpha-phenyl ethynyl-benzenemethanol
PubChem CID 296659
IUPAC Name 1,3-diphenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CC(C2=CC=CC=C2)O

Specifications

Density 1.099
Flash Point >110°C (230°F)
Refractive Index 1.618
Quantity 1 g
UN Number UN3082
Formula Weight 208.26
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,3-Diphenyl-2-propyn-1-ol
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only