missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-2-propanone 97.0+%, TCI America™

Catalog No. D091125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D091125G 25 g
D0911100G 100 g
D0911500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D091125G Supplier TCI America Supplier No. D091125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-04-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00004795
InChI Key YFKBXYGUSOXJGS-UHFFFAOYSA-N
Synonym 1,3-diphenylacetone, dibenzyl ketone, 1,3-diphenyl-2-propanone, benzyl ketone, 2-propanone, 1,3-diphenyl, 1,3-diphenylpropanone, alpha,alpha'-diphenylacetone, 1,3-diphenyl-propan-2-one, unii-9y07g5udkq, fema no. 2397
PubChem CID 7593
IUPAC Name 1,3-diphenylpropan-2-one
SMILES O=C(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Diphenyl-2-propanone