missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diphenyl-2,3-epoxy-1-propanone 98.0+%, TCI America™
$46.60 - $141.67
Chemical Identifiers
| CAS | 5411-12-1 |
|---|---|
| Molecular Formula | C15H12O2 |
| Molecular Weight (g/mol) | 224.26 |
| MDL Number | MFCD00022336 |
| InChI Key | UQGMJZQVDNZRKT-UHFFFAOYNA-N |
| Synonym | 2-benzoyl-3-phenyloxirane, chalcone oxide, 1,3-diphenyl-2,3-epoxy-1-propanone, chalcone epoxide, trans-chalcone oxide, phenyl 3-phenyloxiran-2-yl methanone, ccris 2214, trans-phenyl 3-phenyloxiranyl methanone, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone |
| PubChem CID | 92219 |
| IUPAC Name | phenyl-(3-phenyloxiran-2-yl)methanone |
| SMILES | C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3 |
Chemical Identifiers
| 5411-12-1 | |
| 224.26 | |
| UQGMJZQVDNZRKT-UHFFFAOYNA-N | |
| 92219 | |
| C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3 |
| C15H12O2 | |
| MFCD00022336 | |
| 2-benzoyl-3-phenyloxirane, chalcone oxide, 1,3-diphenyl-2,3-epoxy-1-propanone, chalcone epoxide, trans-chalcone oxide, phenyl 3-phenyloxiran-2-yl methanone, ccris 2214, trans-phenyl 3-phenyloxiranyl methanone, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone | |
| phenyl-(3-phenyloxiran-2-yl)methanone |
Specifications
| 5411-12-1 | |
| White | |
| MFCD00022336 | |
| 2-benzoyl-3-phenyloxirane, chalcone oxide, 1,3-diphenyl-2,3-epoxy-1-propanone, chalcone epoxide, trans-chalcone oxide, phenyl 3-phenyloxiran-2-yl methanone, ccris 2214, trans-phenyl 3-phenyloxiranyl methanone, methanone, phenyl 3-phenyloxiranyl, phenyl 3-phenyl-2-oxiranyl methanone | |
| C1=CC=C(C=C1)C2C(O2)C(=O)C3=CC=CC=C3 | |
| 224.26 | |
| 224.26 | |
| Crystalline Powder |
| 90°C | |
| C15H12O2 | |
| 1 g | |
| UQGMJZQVDNZRKT-UHFFFAOYNA-N | |
| phenyl-(3-phenyloxiran-2-yl)methanone | |
| 92219 | |
| ≥98.0% (GC) | |
| 1,3-Diphenyl-2,3-epoxy-1-propanone |
Safety and Handling
TSCA : No