missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-1-butanone, 95%, Thermo Scientific Chemicals

Catalog No. p-7025262
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2065403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2065403 Supplier Thermo Scientific Chemicals Supplier No. B2065403

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1533-20-6
Molecular Formula C16H16O
Molecular Weight (g/mol) 224.303
MDL Number MFCD00026345
InChI Key GIVFXLVPKFXTCU-UHFFFAOYSA-N
Synonym 1,3-diphenyl-1-butanone, 3-phenylbutyrophenone, b-phenylbutyrophenone, acmc-20ap3x, 1,3-diphenyl-butan-1-one, phenyl 2-phenylpropyl ketone, 1-butanone, 1,3-diphenyl, 2-alpha-methylbenzyl acetophenone
PubChem CID 137065
IUPAC Name 1,3-diphenylbutan-1-one
SMILES CC(CC(=O)C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 68°C to 72°C
Quantity 1 g
Formula Weight 224.3
Percent Purity 95%
Chemical Name or Material 1,3-Diphenyl-1-butanone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only