Learn More
1,3-Diphenyl-1,3-propanedione, 98+%
CAS: 120-46-7 | C15H12O2 | 224.259 g/mol
Supplier: Thermo Scientific Chemicals A1345022
Description
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.
Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
Specifications
| 1,3-Diphenyl-1,3-propanedione | |
| 76°C to 80°C | |
| C15H12O2 | |
| 100 g | |
| 14,3009 | |
| It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water. | |
| C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2 | |
| 224.259 | |
| CHEBI:75417 | |
| ≥98% |
| 120-46-7 | |
| 219°C to 221°C (18 mmHg) | |
| MFCD00003085 | |
| 514910 | |
| dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc | |
| NZZIMKJIVMHWJC-UHFFFAOYSA-N | |
| 1,3-diphenylpropane-1,3-dione | |
| 8433 | |
| 224.26 |
Chemical Identifiers
| 120-46-7 | |
| 224.259 | |
| NZZIMKJIVMHWJC-UHFFFAOYSA-N | |
| 8433 | |
| 1,3-diphenylpropane-1,3-dione |
| C15H12O2 | |
| MFCD00003085 | |
| dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc | |
| CHEBI:75417 | |
| C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2 |
Safety and Handling
EINECSNumber : 204-398-9
RTECSNumber : TZ1930000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only