missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylnaphthalene 95.0+%, TCI America™

Catalog No. D08301G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08301G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D08301G Supplier TCI America Supplier No. D08301G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 575-41-7
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004036
InChI Key QHJMFSMPSZREIF-UHFFFAOYSA-N
Synonym naphthalene, 1,3-dimethyl, 1,3-dmn, 1,3-dimethyl-naphthalene, acmc-1ajsv, 1,3-dimethyl naphthalene, naphthalene,1,3-dimethyl, 4-05-00-01708 beilstein handbook reference, 1,3-dimethylnaphthalene
PubChem CID 11327
ChEBI CHEBI:48614
IUPAC Name 1,3-dimethylnaphthalene
SMILES CC1=CC(C)=C2C=CC=CC2=C1

Specifications

Color Yellow
Boiling Point 263°C
Quantity 1 g
UN Number 3082
Formula Weight 156.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylnaphthalene
EINECSNumber (4)-0080
RTECSNumber QJ4420000
TSCA No