missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylnaphthalene, 96%

Catalog No. AC346900010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC346900010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC346900010 Supplier Thermo Scientific Chemicals Supplier No. 346900010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 575-41-7 | C12H12 | 156.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 575-41-7
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004036
InChI Key QHJMFSMPSZREIF-UHFFFAOYSA-N
Synonym naphthalene, 1,3-dimethyl, 1,3-dmn, 1,3-dimethyl-naphthalene, acmc-1ajsv, 1,3-dimethyl naphthalene, naphthalene,1,3-dimethyl, 4-05-00-01708 beilstein handbook reference, 1,3-dimethylnaphthalene
PubChem CID 11327
ChEBI CHEBI:48614
IUPAC Name 1,3-dimethylnaphthalene
SMILES CC1=CC(C)=C2C=CC=CC2=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point -5°C
Color Yellow
Density 0.9820g/mL
Boiling Point 263°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6080 to 1.61
Linear Formula C10H6(CH3)2
Quantity 1 g
Specific Gravity 0.982
Solubility Information Solubility in water: insoluble in water (20°C)
Formula Weight 156.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1, 3-Dimethylnaphthalene, 96%
Show More Show Less
EINECSNumber 209-384-6

RUO – Research Use Only