missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethylimidazolium Methyl Sulfate 98.0+%, TCI America™
$143.54 - $426.87
Chemical Identifiers
| CAS | 97345-90-9 |
|---|---|
| Molecular Formula | C6H12N2O4S |
| Molecular Weight (g/mol) | 208.232 |
| MDL Number | MFCD03453276 |
| InChI Key | WOKQGMYCUGJNIJ-UHFFFAOYSA-M |
| PubChem CID | 12075598 |
| IUPAC Name | 1,3-dimethylimidazol-1-ium;methyl sulfate |
| SMILES | CN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
Chemical Identifiers
| 97345-90-9 | |
| 208.232 | |
| WOKQGMYCUGJNIJ-UHFFFAOYSA-M | |
| 1,3-dimethylimidazol-1-ium;methyl sulfate |
| C6H12N2O4S | |
| MFCD03453276 | |
| 12075598 | |
| CN1C=C[N+](=C1)C.COS(=O)(=O)[O-] |
Specifications
| 97345-90-9 | |
| MFCD03453276 | |
| 1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc | |
| CN1C=C[N+](=C1)C.COS(=O)(=O)[O-] | |
| 208.232 | |
| 208.23 | |
| Liquid |
| C6H12N2O4S | |
| 5 g | |
| WOKQGMYCUGJNIJ-UHFFFAOYSA-M | |
| 1,3-dimethylimidazol-1-ium;methyl sulfate | |
| 12075598 | |
| ≥98.0% (HPLC,N) | |
| 1,3-Dimethylimidazolium Methyl Sulfate |
Safety and Handling
TSCA : No