missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylimidazolium Dimethyl Phosphate 97.0+%, TCI America™

Catalog No. D32405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32405G 5 g
D324025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32405G Supplier TCI America Supplier No. D32405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 654058-04-5
Molecular Formula C7H15N2O4P
Molecular Weight (g/mol) 222.181
InChI Key GSGLHYXFTXGIAQ-UHFFFAOYSA-M
Synonym 1,3-dimethylimidazolium dimethyl phosphate, acmc-1b6nn, 1,3-dimethylimidazoliumdimethylphosphate, dsstox_cid_27946, dsstox_rid_82698, dsstox_gsid_47970, 1-methyl-3-methylimidazolium dimethylphosphate, 1,3-dimethyl-imidazolium dimethylphosphate, 1,3-dimethylimidazolium dimethyl phosphate hplc
PubChem CID 16069365
IUPAC Name 1,3-dimethylimidazol-1-ium;dimethyl phosphate
SMILES CN1C=C[N+](=C1)C.COP(=O)([O-])OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 222.18
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylimidazolium Dimethyl Phosphate