missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. D53495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53495G 5 g
D534925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53495G Supplier TCI America Supplier No. D53495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 174899-81-1
Molecular Formula C7H9F6N3O4S2
Molecular Weight (g/mol) 377.276
MDL Number MFCD18251322
InChI Key SAYMDKMGIAANGQ-UHFFFAOYSA-N
PubChem CID 11349411
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1,3-dimethylimidazol-1-ium
SMILES CN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 22°C
Color White-Yellow
Quantity 5 g
Formula Weight 377.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3-Dimethylimidazolium Bis(trifluoromethanesulfonyl)imide