missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylcyclohexane (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. D069925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069925ML 25 mL
D0699500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D069925ML Supplier TCI America Supplier No. D069925ML
Only null left

Chemical Identifiers

CAS 591-21-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00064173
InChI Key SGVUHPSBDNVHKL-UHFFFAOYSA-N
Synonym hexahydro-m-xylene, cyclohexane, 1,3-dimethyl, 1,3-dimethylcyclohexane,c&t, trans-1,3-dimethylcyclohexane, 1,3-dimethylcyclohexane, cis + trans, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, m-dimethylcyclohexane, acmc-209nia, 1,3-dimethyl cyclohexane
PubChem CID 11564
IUPAC Name 1,3-dimethylcyclohexane
SMILES CC1CCCC(C1)C

Specifications

Color Colorless
Quantity 25 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylcyclohexane (cis- and trans- mixture)
EINECSNumber (3)-2227
TSCA No