missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylbutylamine 98.0+%, TCI America™

Catalog No. D06945ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06945ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D06945ML Supplier TCI America Supplier No. D06945ML

Chemical Identifiers

CAS 108-09-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00008088
InChI Key UNBMPKNTYKDYCG-UHFFFAOYSA-N
Synonym 1,3-dimethylbutylamine, 2-pentanamine, 4-methyl, 2-amino-4-methylpentane, 1,3-dimethylbutanamine, 4-methyl-2-aminopentane, 1,3-dimethyl butylamine, butylamine, 1,3-dimethyl, ccris 4800, 1,3-dimethylbutyl amine, 1,3-dimethyl-n-butylamine
PubChem CID 7908
IUPAC Name 4-methylpentan-2-amine
SMILES CC(C)CC(C)N

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 mL
UN Number 2379
Formula Weight 101.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylbutylamine
RTECSNumber EO4460000
TSCA Yes