missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 45°C |
| Quantity | 5 g |
| UN Number | 2735 |
| Formula Weight | 155.25 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide |
Safety and Handling
| EINECSNumber | (2)-1696 |
| TSCA | Yes |
| Recommended Storage | Freezer |