missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

Catalog No. AAA1096203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1096203 1 g
AAA1096206 5 g
AAA1096214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1096203 Supplier Thermo Scientific Chemicals Supplier No. A1096203

CAS: 22572-40-3 | C9H20N3+ | 170.28 g/mol

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.

Chemical Identifiers

CAS 22572-40-3
Molecular Formula C9H20N3+
Molecular Weight (g/mol) 170.28
MDL Number MFCD00011833
InChI Key JHMDMCSGQQLHNA-UHFFFAOYSA-N
Synonym 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
PubChem CID 122243
IUPAC Name 3-(ethyliminomethylideneamino)propyl-trimethylazanium
SMILES CCN=C=NCCC[N+](C)(C)C

Specifications

Melting Point 90°C to 92°C
Quantity 1 g
Beilstein 4162760
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 297.17
Percent Purity 98%
Chemical Name or Material 1- (3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide
Hazard Category H302-H315-H317-H318-H334-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 245-095-1
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only