missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminophenyl)ethanol 98.0+%, TCI America™

Catalog No. D12695G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12695G 5 g
D126925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12695G Supplier TCI America Supplier No. D12695G

Chemical Identifiers

CAS 5339-01-5
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00059658
InChI Key XYGWUVVZDWJPBW-UHFFFAOYNA-N
Synonym 3-Dimethylamino-alpha-methylbenzyl Alcohol, 1-(3-Dimethylaminophenyl)ethyl Alcohol
PubChem CID 220552
IUPAC Name 1-[3-(dimethylamino)phenyl]ethanol
SMILES CC(C1=CC(=CC=C1)N(C)C)O

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Dimethylaminophenyl)ethanol