missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide Methiodide 98.0+%, TCI America™
$150.16 - $150.16
Chemical Identifiers
| CAS | 22572-40-3 |
|---|---|
| Molecular Formula | C9H20N3+ |
| Molecular Weight (g/mol) | 170.28 |
| MDL Number | MFCD00011833 |
| InChI Key | JHMDMCSGQQLHNA-UHFFFAOYSA-N |
| Synonym | 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene |
| PubChem CID | 122243 |
| IUPAC Name | 3-(ethyliminomethylideneamino)propyl-trimethylazanium |
| SMILES | CCN=C=NCCC[N+](C)(C)C |
Chemical Identifiers
| 22572-40-3 | |
| 170.28 | |
| JHMDMCSGQQLHNA-UHFFFAOYSA-N | |
| 122243 | |
| CCN=C=NCCC[N+](C)(C)C |
| C9H20N3+ | |
| MFCD00011833 | |
| 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene | |
| 3-(ethyliminomethylideneamino)propyl-trimethylazanium |
Specifications
| 22572-40-3 | |
| White | |
| MFCD00011833 | |
| 3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium, 1-3-trimethylaminio propyl-3-ethylcarbodiimide, 3-ethyliminomethylideneamino propyl-trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 3-ethylimino methylidene amino propyl trimethylazanium, 1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene | |
| CCN=C=NCCC[N+](C)(C)C | |
| 170.28 | |
| 297.18 | |
| Crystalline Powder |
| 97°C | |
| C9H20N3+ | |
| 5 g | |
| JHMDMCSGQQLHNA-UHFFFAOYSA-N | |
| 3-(ethyliminomethylideneamino)propyl-trimethylazanium | |
| 122243 | |
| ≥98.0% (T) | |
| 1-[3-(Dimethylamino)propyl]-3-ethylcarbodiimide Methiodide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator