missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyladamantane 99.0+%, TCI America™
$100.67 - $296.38
Chemical Identifiers
| CAS | 702-79-4 |
|---|---|
| Molecular Formula | C12H20 |
| Molecular Weight (g/mol) | 164.29 |
| MDL Number | MFCD00074755 |
| InChI Key | CWNOIUTVJRWADX-UHFFFAOYSA-N |
| Synonym | adamantane, 1,3-dimethyl, 1-3-dimethyladamantane, 1,3-dimethyl adamantane, unii-96g4od5z59, 1,3-dimethyltricyclo 3.3.1.13,7 decane, 1,3-dimethyltricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl, 1r,3r,5r,7r-1,3-dimethyladamantane, memantine intermediate |
| PubChem CID | 12800 |
| ChEBI | CHEBI:47900 |
| IUPAC Name | 1,3-dimethyladamantane |
| SMILES | CC12CC3CC(C1)CC(C)(C3)C2 |
Chemical Identifiers
| 702-79-4 | |
| 164.29 | |
| CWNOIUTVJRWADX-UHFFFAOYSA-N | |
| 12800 | |
| 1,3-dimethyladamantane |
| C12H20 | |
| MFCD00074755 | |
| adamantane, 1,3-dimethyl, 1-3-dimethyladamantane, 1,3-dimethyl adamantane, unii-96g4od5z59, 1,3-dimethyltricyclo 3.3.1.13,7 decane, 1,3-dimethyltricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl, 1r,3r,5r,7r-1,3-dimethyladamantane, memantine intermediate | |
| CHEBI:47900 | |
| CC12CC3CC(C1)CC(C)(C3)C2 |
Specifications
| 702-79-4 | |
| 201°C | |
| MFCD00074755 | |
| 3295 | |
| CWNOIUTVJRWADX-UHFFFAOYSA-N | |
| 1,3-dimethyladamantane | |
| 12800 | |
| 164.29 | |
| Liquid |
| Colorless | |
| C12H20 | |
| 5 mL | |
| adamantane, 1,3-dimethyl, 1-3-dimethyladamantane, 1,3-dimethyl adamantane, unii-96g4od5z59, 1,3-dimethyltricyclo 3.3.1.13,7 decane, 1,3-dimethyltricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl, 1r,3r,5r,7r-1,3-dimethyladamantane, memantine intermediate | |
| CC12CC3CC(C1)CC(C)(C3)C2 | |
| 164.29 | |
| CHEBI:47900 | |
| ≥99.0% (GC) | |
| 1,3-Dimethyladamantane |
Safety and Handling
TSCA : No