missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyladamantane 99.0+%, TCI America™

Catalog No. D10815ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10815ML 5 mL
D108125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10815ML Supplier TCI America Supplier No. D10815ML

Chemical Identifiers

CAS 702-79-4
Molecular Formula C12H20
Molecular Weight (g/mol) 164.29
MDL Number MFCD00074755
InChI Key CWNOIUTVJRWADX-UHFFFAOYSA-N
Synonym adamantane, 1,3-dimethyl, 1-3-dimethyladamantane, 1,3-dimethyl adamantane, unii-96g4od5z59, 1,3-dimethyltricyclo 3.3.1.13,7 decane, 1,3-dimethyltricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl, 1r,3r,5r,7r-1,3-dimethyladamantane, memantine intermediate
PubChem CID 12800
ChEBI CHEBI:47900
IUPAC Name 1,3-dimethyladamantane
SMILES CC12CC3CC(C1)CC(C)(C3)C2

Specifications

Color Colorless
Boiling Point 201°C
Quantity 5 mL
UN Number 3295
Formula Weight 164.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyladamantane