missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyladamantane, 98%

Catalog No. AAA1968406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1968406 5 g
AAA1968414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1968406 Supplier Thermo Scientific Chemicals Supplier No. A1968406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 702-79-4 | C12H20 | 164.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 702-79-4
Molecular Formula C12H20
Molecular Weight (g/mol) 164.29
MDL Number MFCD00074755
InChI Key CWNOIUTVJRWADX-UHFFFAOYSA-N
Synonym adamantane, 1,3-dimethyl, 1-3-dimethyladamantane, 1,3-dimethyl adamantane, unii-96g4od5z59, 1,3-dimethyltricyclo 3.3.1.13,7 decane, 1,3-dimethyltricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl, 1r,3r,5r,7r-1,3-dimethyladamantane, memantine intermediate
PubChem CID 12800
ChEBI CHEBI:47900
IUPAC Name 1,3-dimethyladamantane
SMILES CC12CC3CC(C1)CC(C)(C3)C2

Specifications

Density 0.902
Boiling Point 201°C
Flash Point 52°C (126°F)
Refractive Index 1.478
Quantity 5 g
UN Number UN3295
Formula Weight 164.29
Percent Purity 98%
Chemical Name or Material 1,3-Dimethyladamantane
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-870-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only