missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-6-chlorouracil 98.0+%, TCI America™

Catalog No. D16195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16195G 5 g
D161925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16195G Supplier TCI America Supplier No. D16195G
Only null left

Chemical Identifiers

CAS 6972-27-6
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.58
MDL Number MFCD00038066
InChI Key VATQPUHLFQHDBD-UHFFFAOYSA-N
Synonym 6-chloro-1,3-dimethyluracil, 6-chloro-1,3-dimethylpyrimidine-2,4 1h,3h-dione, 1,3-dimethyl-6-chlorouracil, 6-chloro-1,3-dimethyl-2,4 1h,3h-pyrimidinedione, 2,4 1h,3h-pyrimidinedione, 6-chloro-1,3-dimethyl, 6-chloro-2,4-dihydroxy-1,3-dimethylpyrimidine, 6-chloro-1,3-dimethyl-2,4-1h,3h-pyrimidinedione, 6-chloro-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione, acmc-209oan, 1,3-dimethyl-4-chlorouracil
PubChem CID 81442
IUPAC Name 6-chloro-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CN1C(Cl)=CC(=O)N(C)C1=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyl-6-chlorouracil