missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-5-pyrazolone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 117°C |
| Quantity | 25 g |
| Formula Weight | 112.13 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3-Dimethyl-5-pyrazolone |
Safety and Handling
| EINECSNumber | (5)-5771 |
| TSCA | Yes |