missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-pyrazolone 98.0+%, TCI America™

Catalog No. D203125G
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D203125G 25 g
D2031100G 100 g
D2031500G 500 g
3 options

Catalog No. D203125G

Supplier: TCI America D203125G

Only null left

Chemical Identifiers

CAS 2749-59-9
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00067008
InChI Key NDELSWXIAJLWOU-UHFFFAOYSA-N
Synonym 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one
PubChem CID 17673
IUPAC Name 2,5-dimethyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 112.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyl-5-pyrazolone
EINECSNumber (5)-5771
TSCA Yes