missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-aminoadamantane hydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 298080100

Quantity 10g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC298080100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

41100-52-1
215.77
LDDHMLJTFXJGPI-UHFFFAOYNA-N
181458
3,5-dimethyladamantan-1-amine;hydrochloride
C12H22ClN
MFCD00214336
memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
CHEBI:64323
[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

1, 3-Dimethyl-5-aminoadamantane hydrochloride
Authentic
Glass bottle
MFCD00214336
15,5897
Solubility in water: soluble
[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2
215.77
CHEBI:64323
99%
41100-52-1
99%
C12H22ClN
10g
memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
LDDHMLJTFXJGPI-UHFFFAOYNA-N
3,5-dimethyladamantan-1-amine;hydrochloride
181458
215.77
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec

GHS Signal Word: Warning

EINECSNumber : 255-219-6

SDS
Documents

Documents

Promotions

Promotions