missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-aminoadamantane hydrochloride, 99%, Thermo Scientific™

Catalog No. AC298080010
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC298080010 1g Glass bottle
AC298080100 10g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC298080010 Supplier Thermo Scientific Chemicals Supplier No. 298080010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 41100-52-1
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00214336
InChI Key LDDHMLJTFXJGPI-UHFFFAOYNA-N
Synonym memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
PubChem CID 181458
ChEBI CHEBI:64323
IUPAC Name 3,5-dimethyladamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1g
Merck Index 15,5897
Solubility Information Solubility in water: soluble
Formula Weight 215.77
Percent Purity 99%
Chemical Name or Material 1, 3-Dimethyl-5-aminoadamantane hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
EINECSNumber 255-219-6

RUO – Research Use Only