missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 97.0+%, TCI America™

Catalog No. D51841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51841G Supplier TCI America Supplier No. D51841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1046832-21-6
Molecular Formula C11H19BN2O2
Molecular Weight (g/mol) 222.095
MDL Number MFCD09864190
InChI Key FBNAMBTYMSWTIB-UHFFFAOYSA-N
Synonym 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3-dimethyl-1h-pyrazole-4-boronic acid,pinacol ester, 1,3-dimethylpyrazole-4-boronic acid pinacol ester, 1,3-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-1,3-dimethylpyrazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1h-pyrazole, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem23961
PubChem CID 42647316
IUPAC Name 1,3-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2C)C

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 1 g
Formula Weight 222.10
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole