missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-thiohydantoin 98.0+%, TCI America™

Catalog No. D25065G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25065G Supplier TCI America Supplier No. D25065G
Only null left

Chemical Identifiers

CAS 1801-62-3
Molecular Formula C5H8N2OS
Molecular Weight (g/mol) 144.192
MDL Number MFCD00186242
InChI Key GKWNLFPHQCFFQJ-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-thiohydantoin, 1,3-dimethyl-2-thioxoimidazolidin-4-one, n,n'-dimethylthiohydantoin, 1,3-dimethyl-2-thioxo-4-imidazolidinone, 4-imidazolidinone,1,3-dimethyl-2-thioxo, acmc-1c9l4, 1,3-dimethyl-2-thioxo-1,3-diazolidin-4-one
PubChem CID 3722203
IUPAC Name 1,3-dimethyl-2-sulfanylideneimidazolidin-4-one
SMILES CN1CC(=O)N(C1=S)C

Specifications

Melting Point 93°C
Quantity 5 g
Formula Weight 144.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyl-2-thiohydantoin