missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-2-imidazolidinone, Solstice

Catalog No. 6002817
Change view
Click to view available options
Quantity:
100 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002817 100 mL
6002818 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002817 Supplier Solstice Supplier No. 67484100ML

CHROMASOLV™, GC-Headspace tested,≥99.5%

Solstice Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests

Chemical Identifiers

CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O

Specifications

Density 1.056g/mL at 25°C
Flash Point 95°C (203°F)
Quantity 100 mL
UN Number NONH for all modes of transport
Beilstein 108808
Formula Weight 114.15g/mol
Percent Purity ≥99.5% (GC), ≥99.5%
Grade CHROMASOLV
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone
Hazard Statement P280-P305 + P351 + P338
RTECSNumber NJ0660000