missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 1,3-Dimethyl-2-imidazolidinone

Catalog No. 40725500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40725500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40725500ML Supplier Sigma Aldrich Supplier No. 40725500ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,3-dimethyl-2-imidazolidinone (dmi) is an imidazolidine derivative. it is reported to act as a promoter by minimizing the formation of dialkylation byproducts and accelerating the rate of monoalkylation of α-butyrolactone. conversion of co2 to form lower alcohols by homogeneous catalytic hydrogenation using dmi as solvent has been investigated.1,3-dimethyl-2-imidazolidinone may be used:
  • As substitute solvent for hmpa in the synthesis of 1,2-bis(trimethylsilyl)benzene
  • As solvent during the α-regioselective prenylation of imines
  • A component of mobile phase during size-exclusion chromatographic analysis of cellulose.solvent used in various synthetic organic transformations. studied in the formation of functionally stabilized hydrosilanediyl-transition metal complexes produced photochemically from arylsilanes

Specifications

CAS 80-73-9
Density 1.056 g/mL (at 25°C (literature))
Boiling Point 224°C to 226°C (lit.)
Molecular Formula C5H10N2O
Linear Formula C5H10N2O
Refractive Index n20/D 1.472 (literature)
MDL Number MFCD00003188
Quantity 500 mL
Synonym N,N-Dimethylethyleneurea; DMEU; DMI
Molecular Weight (g/mol) 114.15
Percent Purity ≥99.5% (GC)
Show More Show Less