missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. p-2694755
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC22902CB 250mg Amber Glass Bottle
CC22902DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC22902CB Supplier Thermo Scientific Chemicals Supplier No. CC22902CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 388088-75-3
Molecular Formula C8H7ClN2OS
Molecular Weight (g/mol) 214.67
MDL Number MFCD03086153
InChI Key BQOTYHLFKDBAGL-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethylthieno 2,3-c pyrazole-5-carbonyl chloride, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carbonyl chloride, 1h-thieno 2,3-c pyrazole-5-carbonylchloride, 1,3-dimethyl
PubChem CID 2776542
IUPAC Name 1,3-dimethylthieno[2,3-c]pyrazole-5-carbonyl chloride
SMILES CN1N=C(C)C2=C1SC(=C2)C(Cl)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 214.68
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carbonyl chloride
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.