missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC12501EA
Change view
Click to view available options
Quantity:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12501EA 10g
CC12501DA 1g
CC12501DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC12501EA Supplier Thermo Scientific Chemicals Supplier No. CC12501EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5744-56-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00159636
InChI Key QRWZFUBHOQWUGH-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl
PubChem CID 587721
IUPAC Name 2,5-dimethylpyrazole-3-carboxylic acid
SMILES CN1N=C(C)C=C1C(O)=O

Specifications

Assay Percent Range 97%
Quantity 10g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid
Hazard Category Warning
Hazard Statement GHS P Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.