missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12501DE
| Quantity | 5g |
|---|
Chemical Identifiers
| C6H8N2O2 | |
| MFCD00159636 | |
| 1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl | |
| 2,5-dimethylpyrazole-3-carboxylic acid |
Specifications
| 1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid | |
| 97% | |
| MFCD00159636 | |
| 1,3-dimethyl-1h-pyrazole-5-carboxylic acid, 1,3-dimethylpyrazole-5-carboxylic acid, 2,5-dimethyl-2h-pyrazole-3-carboxylic acid, 1h-pyrazole-5-carboxylic acid, 1,3-dimethyl, 1,3-dimethylpyrazole-5-carboxylicacid, pubchem7839, ksc269g2l, 1,3-dimethylpyrazol-5-carboxylic acid, pyrazole-5-carboxylic acid, 1,3-dmethyl, 1h-pyrazole-5-carboxylicacid, 1,3-dimethyl | |
| CN1N=C(C)C=C1C(O)=O | |
| 140.14 | |
| 140.14 |
| 5744-56-9 | |
| C6H8N2O2 | |
| 5g | |
| QRWZFUBHOQWUGH-UHFFFAOYSA-N | |
| 2,5-dimethylpyrazole-3-carboxylic acid | |
| 587721 | |
| 97% |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
Warning