missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethoxy-1,3-dimethylurea 97.0+%, TCI America™

Catalog No. d39905g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39905G Supplier TCI America Supplier No. D39905G
Only null left

Chemical Identifiers

CAS 123707-26-6
Molecular Formula C5H12N2O3
Molecular Weight (g/mol) 148.16
MDL Number MFCD22041645
InChI Key BXBKNWRSAAAPKG-UHFFFAOYSA-N
PubChem CID 11159391
IUPAC Name 1,3-dimethoxy-1,3-dimethylurea
SMILES CON(C)C(=O)N(C)OC

Specifications

Color Yellow
Boiling Point 42°C
Quantity 5 g
UN Number 1993
Formula Weight 148.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethoxy-1,3-dimethylurea
TSCA No
Recommended Storage Refrigerator