missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimesitylimidazolium Chloride 98.0+%, TCI America™

Catalog No. D34461G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34461G 1 g
D34465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34461G Supplier TCI America Supplier No. D34461G

Chemical Identifiers

CAS 141556-45-8
Molecular Formula C21H25ClN2
Molecular Weight (g/mol) 340.90
MDL Number MFCD02684541
InChI Key OTOSIXGMLYKKOW-UHFFFAOYSA-M
Synonym 1,3-bis 2,4,6-trimethylphenyl imidazolium chloride, 1,3-dimesityl-1h-imidazol-3-ium chloride, 1,3-dimesitylimidazolium chloride, 1,3-bis mesityl imidazolium chloride, imes.hcl, 1,3-bis 2,4,6-trimethylphenyl imidazoliumchloride, 1,3-2,4,6-trimethylphenyl imidazolium chloride, 1,3-bis-2,4,6-trimethylphenyl-1h-imidazolium chloride, 1,3-bis-2,4,6-trimethyl-phenyl-3h-imidazol-1-ium chloride, 1,3-dihydro-1,3-dimesityl-2h-imidazol-2-ylidene monohydrochloride
PubChem CID 2734211
IUPAC Name 1,3-bis(2,4,6-trimethylphenyl)-3H-1λ⁵-imidazol-1-ylium chloride
SMILES [Cl-].CC1=CC(C)=C(N2C=C[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1

Specifications

Melting Point 352°C
Color White-Yellow
Quantity 1 g
Formula Weight 340.90
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimesitylimidazolium Chloride