missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane, 99%, stabilized

Catalog No. AC218600250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218600250 25 mL Glass bottle
AC218601000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218600250 Supplier Thermo Scientific Chemicals Supplier No. 218600250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 627-31-6 | C3H6I2 | 295.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 627-31-6
Molecular Formula C3H6I2
Molecular Weight (g/mol) 295.88
MDL Number MFCD00001097
InChI Key AAAXMNYUNVCMCJ-UHFFFAOYSA-N
Synonym trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
PubChem CID 12314
IUPAC Name 1,3-diiodopropane
SMILES C(CI)CI

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Pink to Yellow
Density 2.5760g/mL
Boiling Point 111.0°C to 113.0°C (31.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6405 to 1.6425
Linear Formula I(CH2)3I
Quantity 25 mL
Beilstein 01, 115
Specific Gravity 2.576
Solubility Information Solubility in water: insoluble in water.
Stabilizer copper
Formula Weight 295.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Diiodopropane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-993-4
RTECSNumber TY1080000
TSCA TSCA

RUO – Research Use Only