missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxyacetone dimer, 97%

Catalog No. aaa1418930
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1418930 250 g
AAA1418918 50 g
AAA141890B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1418930 Supplier Thermo Scientific Chemicals Supplier No. A1418930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 62147-49-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00051019
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
Synonym 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
PubChem CID 4180364
SMILES OCC(=O)CO

Specifications

Melting Point ∼75°C to 82°C
Quantity 250 g
Beilstein 112910
Merck Index 14,3182
Solubility Information Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin.
IUPAC Name 1,3-dihydroxypropan-2-one
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 1,3-Dihydroxyacetone dimer
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 612-947-2
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only