missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxyacetone dimer, 97%

CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol

Supplier:  Thermo Scientific Chemicals A141890B

 View more versions of this product

Catalog No. AAA141890B

Only null left
Explore available promotions

Description

Description

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

1,3-Dihydroxyacetone dimer
62147-49-3
C3H6O3
112910
1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
RXKJFZQQPQGTFL-UHFFFAOYSA-N
2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
4180364
97%
∼75°C to 82°C
1000 g
MFCD00051019
14,3182
Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin.
OCC(=O)CO
90.08
180.16

Chemical Identifiers

62147-49-3
90.08
RXKJFZQQPQGTFL-UHFFFAOYSA-N
4180364
C3H6O3
MFCD00051019
1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
OCC(=O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 612-947-2

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only