missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoroacetone, 98%

Catalog No. AAH5114503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5114503 1 g
AAH5114506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5114503 Supplier Thermo Scientific Chemicals Supplier No. H5114503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 453-14-5 | C3H4F2O | 94.061 g/mol

1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 453-14-5
Molecular Formula C3H4F2O
Molecular Weight (g/mol) 94.061
MDL Number MFCD07368665
InChI Key HKIPCXRNASWFRU-UHFFFAOYSA-N
Synonym 1,3-difluoroacetone, 2-propanone, 1,3-difluoro, 1,3-difluoro-2-propanone, ch2f 2co, 1,3-bis fluoranyl propan-2-one, fluoromethyl ketone, sym-difluoroacetone, sgqdlibrbrwsp@, 1, 3-difluoroacetone, 1,3-difluoro acetone
PubChem CID 228397
IUPAC Name 1,3-difluoropropan-2-one
SMILES C(C(=O)CF)F

Specifications

Boiling Point 101°C to 102°C
Quantity 1 g
UN Number UN2929
Formula Weight 94.06
Percent Purity 98%
Chemical Name or Material 1,3-Difluoroacetone
Hazard Category H226-H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only