missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethynylbenzene 96.0+%, TCI America™

Catalog No. D24965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24965G 5 g
D24961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24965G Supplier TCI America Supplier No. D24965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1785-61-1
Molecular Formula C10H6
Molecular Weight (g/mol) 126.158
MDL Number MFCD00041686
InChI Key PNXLPYYXCOXPBM-UHFFFAOYSA-N
PubChem CID 123130
IUPAC Name 1,3-diethynylbenzene
SMILES C#CC1=CC(=CC=C1)C#C

Specifications

Melting Point -3°C
Boiling Point 78°C
Quantity 5 g
Formula Weight 126.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diethynylbenzene