missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylbenzene 97.0+%, TCI America™

Catalog No. p-7126237
Change view
Click to view available options
Quantity:
100 mL
25 mL
Catalog No. Quantity
D047825ML 25 mL
D0478100ML 100 mL
2 options

Catalog No. D047825ML

Supplier: TCI America D047825ML

Only null left

Chemical Identifiers

CAS 141-93-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009260
InChI Key AFZZYIJIWUTJFO-UHFFFAOYSA-N
Synonym m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
PubChem CID 8864
IUPAC Name 1,3-diethylbenzene
SMILES CCC1=CC(=CC=C1)CC

Specifications

Melting Point -84°C
Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 2049
Formula Weight 134.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
EINECSNumber (3)-0013&(3)-0060
RTECSNumber CZ5620000
TSCA Yes