missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethylbenzene 97.0+%, TCI America™

Catalog No. D047825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D047825ML 25 mL
D0478100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D047825ML Supplier TCI America Supplier No. D047825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-93-5
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00009260
InChI Key AFZZYIJIWUTJFO-UHFFFAOYSA-N
Synonym m-diethylbenzene, benzene, 1,3-diethyl, benzene, m-diethyl, m-ethylethylbenzene, unii-zm2x7i1g8y, zm2x7i1g8y, ethylated benzene, benzene, ethylated, meta-diethylbenzene, polyethylated benzene
PubChem CID 8864
IUPAC Name 1,3-diethylbenzene
SMILES CCC1=CC(=CC=C1)CC

Specifications

Melting Point -84°C
Color Yellow
Boiling Point 182°C
Quantity 25 mL
UN Number 2049
Formula Weight 134.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diethylbenzene
EINECSNumber (3)-0013&(3)-0060
RTECSNumber CZ5620000
TSCA Yes